ENA069848622 Enamine

6-(1-chloroethyl)-N-phenyl-1,3,5-triazine-2,4-diamine

  • MDL number MFCD07850243
  • Cas Number : 99860-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23912
Availability details for
Quantity
  • Availability
  • Add to Cart