ENA069848625 Enamine

3-amino-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD07847844
  • Cas Number : 877825-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23917
Availability details for
Quantity
  • Availability
  • Add to Cart