ENA069848632 Enamine

2-(6,7-dimethoxy-4-oxo-3,4-dihydroquinazolin-3-yl)acetic acid

  • MDL number MFCD07847633
  • Cas Number : 877140-09-5
  • Molecular Weight : 264.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23926
Availability details for
Quantity
  • Availability
  • Add to Cart