ENA069848637 Enamine

4-chloro-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD07850245
  • Cas Number : 877964-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23931
Availability details for
Quantity
  • Availability
  • Add to Cart