ENA069848658 Enamine

2-(5-phenyl-2H-tetrazol-2-yl)ethanesulfonyl fluoride

  • MDL number MFCD07850238
  • Cas Number : 877964-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23958

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23958
Availability details for
Quantity
  • Availability
  • Add to Cart