ENA069848663 Enamine

2-(chloromethyl)-5,6-dimethylthieno[2,3-d]pyrimidin-4-amine

  • MDL number MFCD07762675
  • Cas Number : 877964-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23963

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23963
Availability details for
Quantity
  • Availability
  • Add to Cart