ENA069848664 Enamine

2-(N-methyl2,3-dihydro-1,4-benzodioxine-6-sulfonamido)acetic acid

  • MDL number MFCD02225806
  • Cas Number : 425659-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23964
Availability details for
Quantity
  • Availability
  • Add to Cart