ENA069848672 Enamine

thieno[3,2-d]pyrimidin-4(3H)-one

  • MDL number MFCD00122140
  • Cas Number : 16234-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23988

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23988
Availability details for
Quantity
  • Availability
  • Add to Cart