ENA069848680 Enamine

1-[2-(aminomethyl)phenyl]-N-methylmethanesulfonamide

  • MDL number MFCD07847869
  • Cas Number : 877825-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24003
Availability details for
Quantity
  • Availability
  • Add to Cart