ENA069848694 Enamine

2-mercapto-3-methylthieno[3,2-d]pyrimidin-4(3H)-one

  • MDL number MFCD07850257
  • Cas Number : 183170-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 313...315
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24027

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24027
Availability details for
Quantity
  • Availability
  • Add to Cart