ENA069848718 Enamine

(4-chloro-2-methyl-3,5-dinitrophenyl)methanol

  • MDL number MFCD07852067
  • Cas Number : 72908-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24067
Availability details for
Quantity
  • Availability
  • Add to Cart