ENA069848729 Enamine

2-(2-oxo-1,2-dihydroquinoxalin-1-yl)ethane-1-sulfonyl fluoride

  • MDL number MFCD07852061
  • Cas Number : 877977-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24082
Availability details for
Quantity
  • Availability
  • Add to Cart