ENA069848739 Enamine

4-hydroxy-5-methoxy-2-nitrobenzaldehyde

  • MDL number MFCD00034536
  • Cas Number : 2454-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-210
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24100
Availability details for
Quantity
  • Availability
  • Add to Cart