ENA069848741 Enamine

2-chloro-1-[4-(3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl)piperazin-1-yl]ethan-1-one

  • MDL number MFCD07850223
  • Cas Number : 877963-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-22949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22949
Availability details for
Quantity
  • Availability
  • Add to Cart