ENA069848748 Enamine

2-chloro-N-(1,1-dioxo-1lambda6-thiolan-3-yl)-N-(2-methoxyethyl)acetamide

  • MDL number MFCD07440312
  • Cas Number : 877964-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23020
Availability details for
Quantity
  • Availability
  • Add to Cart