ENA069848757 Enamine

N-[4-(2-amino-1,3-thiazol-4-yl)-3-fluorophenyl]acetamide

  • MDL number MFCD08239627
  • Cas Number : 878218-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24124
Availability details for
Quantity
  • Availability
  • Add to Cart