ENA069848759 Enamine

3-methylbenzene-1-sulfonamide

  • MDL number MFCD04066695
  • Cas Number : 1899-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-109
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24126

Safety Information

signalword Warning
pcodes P201,P202,P261,P264,P270,P271,P280,P281,P301+P312,P302+P352,P304+P340,P305+P351+P338,P308+P313,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-24126
pictograms GHS07,GHS08
hstatements H302,H315,H319,H335,H351
Availability details for
Quantity
  • Availability
  • Add to Cart