ENA069848776 Enamine

2-chloro-N-[(4-fluorophenyl)(thiophen-2-yl)methyl]acetamide

  • MDL number MFCD07838473
  • Cas Number : 875160-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23239
Availability details for
Quantity
  • Availability
  • Add to Cart