ENA069848786 Enamine

1-(2-methoxyethyl)-6-oxo-1,6-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD07838420
  • Cas Number : 875163-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23417
Availability details for
Quantity
  • Availability
  • Add to Cart