ENA069848792 Enamine

1-(4-methoxy-1-naphthoyl)piperidine-3-carboxylic acid

  • MDL number MFCD07838484
  • Cas Number : 875160-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255...257
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23462

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23462
Availability details for
Quantity
  • Availability
  • Add to Cart