ENA069848797 Enamine

2-(2,3-dioxo-2,3-dihydro-1H-indol-1-yl)-N-(3-methylphenyl)acetamide

  • MDL number MFCD00401994
  • Cas Number : 301819-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15539

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15539
Availability details for
Quantity
  • Availability
  • Add to Cart