ENA087844701 Enamine

2-chloro-N-methyl-N-{[4-(methylsulfanyl)phenyl]methyl}acetamide

  • MDL number MFCD08271731
  • Cas Number : 923695-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-22895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22895
Availability details for
Quantity
  • Availability
  • Add to Cart