ENA087844710 Enamine

4-(2-chloroacetamido)-N-(3-sulfamoylphenyl)benzamide

  • MDL number MFCD08271752
  • Cas Number : 923897-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23224
Availability details for
Quantity
  • Availability
  • Add to Cart