ENA087844725 Enamine

5-(bromomethyl)-3-(2-methoxyphenyl)-1,2-oxazole

  • MDL number MFCD06738512
  • Cas Number : 886364-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24229
Availability details for
Quantity
  • Availability
  • Add to Cart