ENA087844751 Enamine

1-[2-(thiophen-2-yl)ethyl]-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD08444258
  • Cas Number : 923161-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24435
Availability details for
Quantity
  • Availability
  • Add to Cart