ENA087844752 Enamine

2,2,2-trifluoroethyl morpholine-4-carboxylate

  • MDL number MFCD08444260
  • Cas Number : 923177-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24442

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24442
Availability details for
Quantity
  • Availability
  • Add to Cart