ENA087844753 Enamine

1-(1-methyl-1H-pyrrole-2-carbonyl)piperazine

  • MDL number MFCD08444261
  • Cas Number : 473733-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24443
Availability details for
Quantity
  • Availability
  • Add to Cart