ENA087844754 Enamine

(2Z)-2-cyano-N-(4-isopropylphenyl)-2-(4-oxo-3-phenyl-1,3-thiazolidin-2-ylidene)acetamide

  • MDL number MFCD08444262
  • Cas Number : 924869-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24449

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24449
Availability details for
Quantity
  • Availability
  • Add to Cart