ENA087844757 Enamine

1-(2,5-dimethylbenzenesulfonyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD08444267
  • Cas Number : 923221-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24478
Availability details for
Quantity
  • Availability
  • Add to Cart