ENA087844758 Enamine

2-amino-5-methoxy-4-(2-methoxyethoxy)benzoic acid

  • MDL number MFCD08444268
  • Cas Number : 923200-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24479
Availability details for
Quantity
  • Availability
  • Add to Cart