ENA087844759 Enamine

3-(cyclopropylsulfamoyl)thiophene-2-carboxylic acid

  • MDL number MFCD08444271
  • Cas Number : 923129-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24486
Availability details for
Quantity
  • Availability
  • Add to Cart