ENA087844763 Enamine

2-(2-chloroacetyl)-5-methyl-2,3-dihydro-1,2-oxazol-3-one

  • MDL number MFCD08444275
  • Cas Number : 38100-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24492
Availability details for
Quantity
  • Availability
  • Add to Cart