ENA087844768 Enamine

5-(2-nitrophenyl)thiophene-2-carbaldehyde

  • MDL number MFCD08444285
  • Cas Number : 42545-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24518

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24518
Availability details for
Quantity
  • Availability
  • Add to Cart