ENA087844775 Enamine

3-methyl-6-(propan-2-yl)-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08444312
  • Cas Number : 923690-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24702
Availability details for
Quantity
  • Availability
  • Add to Cart