ENA087844801 Enamine

N-[(benzylamino)carbonyl]-2-chloropropanamide

  • MDL number MFCD08262828
  • Cas Number : 879361-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23406

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23406
Availability details for
Quantity
  • Availability
  • Add to Cart