ENA087844803 Enamine

2-chloro-N-[(isopropylamino)carbonyl]propanamide

  • MDL number MFCD08261055
  • Cas Number : 879319-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23409

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23409
Availability details for
Quantity
  • Availability
  • Add to Cart