ENA087844804 Enamine

2-(2-chloroacetamido)-N-[(furan-2-yl)methyl]benzamide

  • MDL number MFCD08245276
  • Cas Number : 117459-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23428
Availability details for
Quantity
  • Availability
  • Add to Cart