ENA087844806 Enamine

N-[4-(2-chloroacetamido)-2,5-dimethoxyphenyl]benzamide

  • MDL number MFCD01252281
  • Cas Number : 289655-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23456
Availability details for
Quantity
  • Availability
  • Add to Cart