ENA087844807 Enamine

2-chloro-N-[1-(furan-2-yl)ethyl]-N-(pyridin-2-yl)acetamide

  • MDL number MFCD03150770
  • Cas Number : 380432-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01828
Availability details for
Quantity
  • Availability
  • Add to Cart