ENA087844808 Enamine

1-(3-methylphenyl)piperazine

  • MDL number MFCD00040734
  • Cas Number : 41186-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...173
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23498

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-23498
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart