ENA087844821 Enamine

[6-(4-fluorophenyl)imidazo[2,1-b][1,3]thiazol-3-yl]acetic acid

  • MDL number MFCD06761607
  • Cas Number : 878259-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24121

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24121
Availability details for
Quantity
  • Availability
  • Add to Cart