ENA087844822 Enamine

2-[(1-methyl-4-nitro-1H-pyrrol-2-yl)formamido]acetic acid

  • MDL number MFCD08246135
  • Cas Number : 98488-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24132
Availability details for
Quantity
  • Availability
  • Add to Cart