ENA087844850 Enamine

N-[2-(2-amino-1,3-thiazol-4-yl)-5-fluorophenyl]acetamide

  • MDL number MFCD08246131
  • Cas Number : 878259-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-128
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24182
Availability details for
Quantity
  • Availability
  • Add to Cart