ENA087844868 Enamine

3-(3-ethyl-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)propanoic acid

  • MDL number MFCD08246115
  • Cas Number : 878259-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24209

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24209
Availability details for
Quantity
  • Availability
  • Add to Cart