ENA087844872 Enamine

1-[(furan-2-yl)methyl]-5-(trifluoromethyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD08246113
  • Cas Number : 878259-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24216
Availability details for
Quantity
  • Availability
  • Add to Cart