ENA087844877 Enamine

2-(morpholin-4-ylcarbonyl)phenol

  • MDL number MFCD00038032
  • Cas Number : 3202-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24222

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24222
Availability details for
Quantity
  • Availability
  • Add to Cart