ENA087844880 Enamine

3,5-dimethyl-1-phenyl-1H-pyrazol-4-amine

  • MDL number MFCD00233463
  • Cas Number : 21683-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-63
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24225
Availability details for
Quantity
  • Availability
  • Add to Cart