ENA087844889 Enamine

2-cyano-N-(3-isopropoxypropyl)acetamide

  • MDL number MFCD08256161
  • Cas Number : 15029-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24234

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24234
Availability details for
Quantity
  • Availability
  • Add to Cart