ENA087844891 Enamine

2-cyano-N-(4-methoxybenzyl)acetamide

  • MDL number MFCD01418247
  • Cas Number : 288154-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24236

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24236
Availability details for
Quantity
  • Availability
  • Add to Cart