ENA087844896 Enamine

2-chloro-N-methyl-N-[(thiophen-3-yl)methyl]acetamide

  • MDL number MFCD08245285
  • Cas Number : 878466-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-22639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22639
Availability details for
Quantity
  • Availability
  • Add to Cart